Learn More
Chemglass Life Sciences Diameter x Height (mm): 125 x 69, Reactor Size: 2L-5L
SDP
Catalog No. CG2081A02
Encompass_Preferred
Diameter x Height (mm): 125 x 69, Reactor Size: 2L-5L. Heavy duty PTFE anchor made of 0.500 thick PTFE. Blade is shaped to mirror the contour of Chemglass cylindrical reactors. Designed for use with CG-2078 Chem-Stir 10mm stirrer shaft. The blade is secured to the stirrer via a PTFE pin. PLEASE NOTE: CG-2081-A-02 IS NOT RECOMMENDED FOR USE WITH BAFFLES OR THERMOCOUPLES.
Catalog No. CG2081A02 Supplier Chemglass Life Sciences Supplier No. CG2081A02
Diameter x Height (mm): 125 x 69, Reactor Size: 2L-5L. Heavy duty PTFE anchor made of 0.500 thick PTFE. Blade is shaped to mirror the contour of Chemglass cylindrical reactors. Designed for use with CG-2078 Chem-Stir 10mm stirrer shaft. The blade is secured to the stirrer via a PTFE pin. PLEASE NOTE: CG-2081-A-02 IS NOT RECOMMENDED FOR USE WITH BAFFLES OR THERMOCOUPLES.