Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ancymidol 98.0+%, TCI America™
SDP

Catalog No. A287725MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A287725MG 25 mg
A2877100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A287725MG Supplier TCI America Supplier No. A287725MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12771-68-5
Molecular Formula C15H16N2O2
Molecular Weight (g/mol) 256.31
MDL Number MFCD00072501
InChI Key HUTDUHSNJYTCAR-UHFFFAOYNA-N
Synonym alpha-Cyclopropyl-alpha-(4-methoxyphenyl)-5-pyrimidinemethanol
PubChem CID 25572
ChEBI CHEBI:73171
IUPAC Name cyclopropyl(4-methoxyphenyl)(pyrimidin-5-yl)methanol
SMILES COC1=CC=C(C=C1)C(O)(C1CC1)C1=CN=CN=C1

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 25 mg
Formula Weight 256.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ancymidol
RTECSNumber UV9280000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.