Learn More
Bioworld Anderson's Rhododendron Medium w/ Vitamins, 50 L
Catalog No. 50254548
Encompass_Preferred
Anderson's Rhododendron MediumSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-548 Supplier Bioworld Supplier No. 306300003
Anderson's Rhododendron MediumSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumPlant tissue culture tested