Learn More
Bioworld Anderson's Rhododendron Medium, 25 L
Catalog No. 50254550
Encompass_Preferred
Anderson's Rhododendron MediumContains Macronutrients and Micronutrients as described by Anderson (1978, 1980)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumWorking Concentration: 1.89 g/LPlant tissue culture tested
Catalog No. 50-254-550 Supplier Bioworld Supplier No. 306300012
Anderson's Rhododendron MediumContains Macronutrients and Micronutrients as described by Anderson (1978, 1980)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of mediumWorking Concentration: 1.89 g/LPlant tissue culture tested