Learn More
Accuflow Systems Anderson-Negele Digital Thermometer, Deg C, 1/2 in. Tri-Clamp, 1 ea
Catalog No. NC3442239
Encompass_Preferred
The FJ Digital Pharma Temperature Sensor from Anderson-Negele provides a reliable and cost-effective solution for process temperature monitoring. It features a large LCD display for clear readability and utilizes field-replaceable batteries, offering a modern alternative to mechanical thermometers.
  • Operates on field-replaceable batteries
  • Large Liquid Crystal Display for easy viewing
  • Field calibration capability for all models
  • Pharma-compliant design with all-welded construction and electro-polished wetted parts
  • High accuracy temperature monitoring with 0.1°C resolution
  • All 316L electropolished wetted parts
Catalog No. NC3442239 Supplier Accuflow Systems Supplier No. FJ0110010110000
The FJ Digital Pharma Temperature Sensor from Anderson-Negele provides a reliable and cost-effective solution for process temperature monitoring. It features a large LCD display for clear readability and utilizes field-replaceable batteries, offering a modern alternative to mechanical thermometers.
  • Operates on field-replaceable batteries
  • Large Liquid Crystal Display for easy viewing
  • Field calibration capability for all models
  • Pharma-compliant design with all-welded construction and electro-polished wetted parts
  • High accuracy temperature monitoring with 0.1°C resolution
  • All 316L electropolished wetted parts