Learn More
Waters Corp Applications Kit, 24-Sample, GlycoWorks Eco, N-Glycan Sample Preparation
Catalog No. 502886195 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for automation-friendly sample preparation of N-linked glycans using an automated robotic system. It modifies starting sample and reagent concentrations to improve pipetting accuracy for volumes of 10 μL or more. It also replaces DMF with DMSO, offering a greener, safer alternative for reagent labeling and removing other DMF-containing solutions.
  • Optimized for automated robotic systems and specific labware.
  • Enhanced pipetting accuracy for volumes ≥10 μL.
  • Eco-friendly reagents, utilizing DMSO instead of DMF.
  • Processes 24 samples in a 3x8 format.
  • Enables fast enzymatic release and rapid labeling of N-glycans.
  • Streamlined protocol minimizes sample handling.
  • Features a novel rapid label for sensitive fluorescence and strong mass detection.
  • Clean-up via solid-phase extraction (SPE) with a HILIC μElution™ Plate.
Catalog No. 50-288-6195 Supplier Waters Corp Supplier No. 176005294
This kit is designed for automation-friendly sample preparation of N-linked glycans using an automated robotic system. It modifies starting sample and reagent concentrations to improve pipetting accuracy for volumes of 10 μL or more. It also replaces DMF with DMSO, offering a greener, safer alternative for reagent labeling and removing other DMF-containing solutions.
  • Optimized for automated robotic systems and specific labware.
  • Enhanced pipetting accuracy for volumes ≥10 μL.
  • Eco-friendly reagents, utilizing DMSO instead of DMF.
  • Processes 24 samples in a 3x8 format.
  • Enables fast enzymatic release and rapid labeling of N-glycans.
  • Streamlined protocol minimizes sample handling.
  • Features a novel rapid label for sensitive fluorescence and strong mass detection.
  • Clean-up via solid-phase extraction (SPE) with a HILIC μElution™ Plate.