Learn More
Waters Corp Application Kit, Andrew+ 96-Sample GlycoWorks Eco, Automation Friendly
Catalog No. 502886227 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Andrew+ 96-Sample GlycoWorks Eco Application Kit automates and streamlines N-linked glycan sample preparation. Designed for the Andrew+ pipetting robot, it provides a quality control and automation-friendly workflow. The kit enhances lab safety by utilizing greener alternatives like DMSO, replacing traditional solvents.
  • Quality control and automation-friendly sample preparation method.
  • Adaptation of standard variable volume method with modified concentrations for improved accuracy.
  • Uses DMSO as a greener and safer alternative to DMF.
  • Processes 96 samples in a 4x24 format.
  • Designed for released N-linked glycan analysis.
  • Compatible with UPLC/UHPLC/HPLC separation modes.
  • Supports HRMS, MS, MS/MS, and UV detection methods.
  • Includes components for deglycosylation, RapiFluor-MS Eco labeling, and HILIC μElution clean-up.
Catalog No. 50-288-6227 Supplier Waters Corp Supplier No. 176005295
Add to Cart
Add to Cart
Request a Quote
This Andrew+ 96-Sample GlycoWorks Eco Application Kit automates and streamlines N-linked glycan sample preparation. Designed for the Andrew+ pipetting robot, it provides a quality control and automation-friendly workflow. The kit enhances lab safety by utilizing greener alternatives like DMSO, replacing traditional solvents.
  • Quality control and automation-friendly sample preparation method.
  • Adaptation of standard variable volume method with modified concentrations for improved accuracy.
  • Uses DMSO as a greener and safer alternative to DMF.
  • Processes 96 samples in a 4x24 format.
  • Designed for released N-linked glycan analysis.
  • Compatible with UPLC/UHPLC/HPLC separation modes.
  • Supports HRMS, MS, MS/MS, and UV detection methods.
  • Includes components for deglycosylation, RapiFluor-MS Eco labeling, and HILIC μElution clean-up.