Learn More
Waters Corp Application Kit, 96HT-Sample, GlycoWorks, Eco, Automated
Catalog No. 502886228 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This application kit offers an eco-friendly, automation-friendly method for high-throughput N-glycan analysis. Validated for use with the Andrew+ robot, it ensures reliable results by adapting variable volume methods for precise pipetting of 10 μL or more. It replaces DMF with DMSO as a greener, safer reagent solvent.
  • Automation-friendly sample preparation for N-glycan analysis.
  • Validated for use with Andrew+ robot.
  • Precise pipetting with 10 μL or more volumes.
  • Uses eco-friendly DMSO solvent.
  • Processes 96 samples in a 2x48 format.
  • Supports UPLC/UHPLC/HPLC and multiple detection methods.
  • Streamlined workflow for rapid processing.
Catalog No. 50-288-6228 Supplier Waters Corp Supplier No. 176005296
Add to Cart
Add to Cart
Request a Quote
This application kit offers an eco-friendly, automation-friendly method for high-throughput N-glycan analysis. Validated for use with the Andrew+ robot, it ensures reliable results by adapting variable volume methods for precise pipetting of 10 μL or more. It replaces DMF with DMSO as a greener, safer reagent solvent.
  • Automation-friendly sample preparation for N-glycan analysis.
  • Validated for use with Andrew+ robot.
  • Precise pipetting with 10 μL or more volumes.
  • Uses eco-friendly DMSO solvent.
  • Processes 96 samples in a 2x48 format.
  • Supports UPLC/UHPLC/HPLC and multiple detection methods.
  • Streamlined workflow for rapid processing.