Learn More
Waters Corp WATERS CORPORATION OligoWorks Protocol, Automates Reagent Addition, 96 Samples, μElution Plate
Catalog No. 502886252 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This OneLab Cloud Library protocol automates the OligoWorks WAX SPE kit for μElution plates, providing maximum flexibility. It streamlines sample preparation and digestion prior to SPE by automating sequential addition of Guanidine, TCEP, and Proteinase K to up to 96 samples in a μElution plate. The Andrew+ robot, using the Microplate Gripper and Heater-Shaker+, ensures consistent results with only one manual capping step before incubation.
  • Automates the sequential addition of Guanidine, TCEP, and Proteinase K.
  • Processes up to 96 samples in a μElution plate.
  • Utilizes the Andrew+ robot, Microplate Gripper tool, and Heater-Shaker+ device.
  • Designed for sample preparation and digestion prior to solid phase extraction (SPE).
  • Requires only one manual step (capping the digestion plate) before incubation.
Catalog No. 50-288-6252 Supplier Waters Corp Supplier No. 176005357
Add to Cart
Add to Cart
Request a Quote
This OneLab Cloud Library protocol automates the OligoWorks WAX SPE kit for μElution plates, providing maximum flexibility. It streamlines sample preparation and digestion prior to SPE by automating sequential addition of Guanidine, TCEP, and Proteinase K to up to 96 samples in a μElution plate. The Andrew+ robot, using the Microplate Gripper and Heater-Shaker+, ensures consistent results with only one manual capping step before incubation.
  • Automates the sequential addition of Guanidine, TCEP, and Proteinase K.
  • Processes up to 96 samples in a μElution plate.
  • Utilizes the Andrew+ robot, Microplate Gripper tool, and Heater-Shaker+ device.
  • Designed for sample preparation and digestion prior to solid phase extraction (SPE).
  • Requires only one manual step (capping the digestion plate) before incubation.