Learn More
Waters Corp OligoWorks Application Kit, Andrew+, Pretreatment/Digestion, SPE Protocols, Bundled
Catalog No. 502886221 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This bundled configuration provides the necessary tools and devices for automating both oligonucleotide sample pretreatment and digestion protocols, as well as SPE protocols using an OligoWorks SPE Microplate Kit.
  • Heater-shaker device with deepwell plate adaptor
  • Extraction base kit with gripper
  • Collection plate spacer kit
  • 8-channel 10-300 μL pipette
  • 1-channel 5-120 μL pipette
  • 8-channel reservoir units
  • Deepwell microplate units
  • Tip insertion system units
Catalog No. 50-288-6221 Supplier Waters Corp Supplier No. 176005359
Add to Cart
Add to Cart
Request a Quote
This bundled configuration provides the necessary tools and devices for automating both oligonucleotide sample pretreatment and digestion protocols, as well as SPE protocols using an OligoWorks SPE Microplate Kit.
  • Heater-shaker device with deepwell plate adaptor
  • Extraction base kit with gripper
  • Collection plate spacer kit
  • 8-channel 10-300 μL pipette
  • 1-channel 5-120 μL pipette
  • 8-channel reservoir units
  • Deepwell microplate units
  • Tip insertion system units