Learn More
Enzo Life Sciences Androgen receptor monoclonal antibody (SPM335) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7700100
Encompass_Preferred
Clone: SPM335. Isotype: IgG1κ. Immunogen: Human androgen receptor, amino acids 302-318 (STEDTAEYSPFKGGYTK). Species reactivity: Human. Specificity: Recognizes a protein of 110kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. Applications: IHC. Purity: Protein G affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-3309