Learn More
Diagnostic Biosystems ANDROGEN RECEPTOR Androgen Receptor
Catalog No. 502407393
Encompass_Preferred
This antibody is specific to a protein of 110 kDa, identified as androgen receptor. This antibody reacts with full length AR and also with the newly described A form of the receptor. This antibody does not cross-react with estrogen, progesterone or glucocorticoid receptors.
Catalog No. 50-240-7393 Supplier Diagnostic Biosystems Supplier No. PDM167
This antibody is specific to a protein of 110 kDa, identified as androgen receptor. This antibody reacts with full length AR and also with the newly described A form of the receptor. This antibody does not cross-react with estrogen, progesterone or glucocorticoid receptors.