Learn More
Biogenex Laboratories Androgen Receptor : 6mL
SDP
Supplier:  Biogenex Laboratories AM2565M
Encompass_Preferred
This monoclonal antibody is specifically designed to recognize a uniqueimmunogenic N-terminal transactivation domain of the androgen receptorthat has a low degree of homology with other steroid receptors. Thisantibody binds to synthetic peptide SP61 of human androgen receptor asassessed by ELISA and recognizes a 100 kD human androgen receptorprotein in Western blot analysis. In addition, this antibody binds to nativehuman androgen receptor in immunoprecipitation, sucrose gradientcentrifugation and immunohistochemistry. This antibody does not crossreact with human estrogen, progesterone or glucocorticoid receptor.
Catalog No. 50-197-8829