Learn More
Medchemexpress LLC HY-130992 5mg Medchemexpress, Androgen receptor antagonist 1 CAS:1338812-36-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1309925MG
Encompass_Preferred
Medchemexpress, HY-130992 5mg Androgen receptor antagonist 1 CAS:1338812-36-4 Androgen receptor antagonist 1 is an orally available full androgen receptor (AR) antagonist with an IC50 of 59 nM. Androgen receptor antagonist 1 (Compound 6) can be used in the synthesis of PROTAC AR degraders, which results 24% and 47 % AR protein degradation in LNCaP cells at 1 μM and 10 μM, respectively[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0244