Learn More
Medchemexpress LLC Androgen receptor H 10ug
SDP
Supplier:  Medchemexpress LLC HYP7208810UG
Encompass_Preferred
Recombinant human androgen receptor protein expressed in E. coli, supplied with N-terminal 10xHis and SUMO tags and a C-terminal Myc tag. It acts as a ligand-activated transcription factor and is provided in microgram quantities for biochemical and molecular studies of androgen receptor signaling.
  • Tagged for affinity purification and detection.
  • High purity greater than 90% by reducing SDS-PAGE.
  • Approximately 58 kDa apparent molecular weight.
  • Expressed in E. coli for scalable production.
  • Stable when stored at -20 °C; aliquot and freeze for long-term storage.
  • Datasheet, COA, and SDS available for quality documentation.
Catalog No. 50-001-96306