Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Androgen receptor H 5ug
SDP

Catalog No. 5000195757
Encompass_Preferred

Recombinant partial fragment of the human androgen receptor (residues D551-T919) expressed in E. coli and supplied as a lyophilized powder with N-terminal His-SUMO and C-terminal Myc tags. The product is formulated in PBS (pH 7.4) with 5 mM DTT, exhibits >90% purity by reducing SDS-PAGE, has endotoxin <1 EU/μg, and is intended for biochemical and cell-based assays evaluating AR activity.

  • Contains AR fragment corresponding to residues D551-T919.
  • Expressed in E. coli and supplied as lyophilized powder.
  • N-terminal His-SUMO and C-terminal Myc tags for detection and purification.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in PBS with 5 mM DTT; recommended storage at -20°C.
  • Demonstrated biological activity with reported ED50 for inhibition of PC-3 cell migration.

Catalog No. 50-001-95757 Supplier Medchemexpress LLC Supplier No. HYP720885UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant partial fragment of the human androgen receptor (residues D551-T919) expressed in E. coli and supplied as a lyophilized powder with N-terminal His-SUMO and C-terminal Myc tags. The product is formulated in PBS (pH 7.4) with 5 mM DTT, exhibits >90% purity by reducing SDS-PAGE, has endotoxin <1 EU/μg, and is intended for biochemical and cell-based assays evaluating AR activity.

  • Contains AR fragment corresponding to residues D551-T919.
  • Expressed in E. coli and supplied as lyophilized powder.
  • N-terminal His-SUMO and C-terminal Myc tags for detection and purification.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in PBS with 5 mM DTT; recommended storage at -20°C.
  • Demonstrated biological activity with reported ED50 for inhibition of PC-3 cell migration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.