Learn More
Medchemexpress LLC Androgen receptor ligand 3 | 693227-00-8 | 99.5% | 343.42 | 500 MG
SDP
Catalog No. 5000404197
Encompass_Preferred
Androgen receptor ligand 3 is an androgen receptor (AR) ligand. It is linked to an IAP ligand via a linker to form an ERα PROTAC degrader.
  • For research use only
  • Not sold to patients
  • White to off-white solid appearance
  • Ships at room temperature in continental US
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-04197 Supplier Medchemexpress LLC Supplier No. HY43962500MG
Androgen receptor ligand 3 is an androgen receptor (AR) ligand. It is linked to an IAP ligand via a linker to form an ERα PROTAC degrader.
  • For research use only
  • Not sold to patients
  • White to off-white solid appearance
  • Ships at room temperature in continental US
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month