Learn More
Medchemexpress LLC Androgen receptor ligand 3 | 693227-00-8 | 99.54% | 343.42 | 100 MG
SDP
Catalog No. 5000403601
Encompass_Preferred
Androgen receptor ligand 3 is an androgen receptor (AR) ligand. It is linked to an IAP ligand via a linker to form an ERα PROTAC degrader.
  • Androgen receptor (AR) ligand
  • Forms an ERα PROTAC degrader
  • High purity of 99.54%
  • Molecular weight of 343.42
  • White to off-white solid appearance
  • For research use only
Catalog No. 50-004-03601 Supplier Medchemexpress LLC Supplier No. HY43962100MG
Androgen receptor ligand 3 is an androgen receptor (AR) ligand. It is linked to an IAP ligand via a linker to form an ERα PROTAC degrader.
  • Androgen receptor (AR) ligand
  • Forms an ERα PROTAC degrader
  • High purity of 99.54%
  • Molecular weight of 343.42
  • White to off-white solid appearance
  • For research use only