Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Androgen Receptor, Ligand Binding Domain, GST Tag, His Tag Recombinant Protein

Catalog No. A15675
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15675 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A15675 Supplier Invitrogen™ Supplier No. A15675
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Androgen receptor ligand binding domain (AR-LBD) is a 64.42 kDa recombinant rat protein, expressed as a GST- and His-tagged protein in insect cells. The protein contains the hinge region and an LBD amino acid sequence identical to that of the human LBD. Differences in the rat and human hinge region are present at human amino acids 646, 649, 650, 655, 656, 657, and 660. AR-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand-binding and coregulator recruitment studies. AR-LBD rat recombinant protein is: Flexible—use in your in-house assay or as part of our PolarScreen™ Competitor assays for a complete assay solution Ready to use—active and tagged (with GST/His) Convenient—available in bulk sizes Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond. Protein Form: Ligand Binding Domain, Recombinant.

The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis.AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain.Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy's disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15207
Concentration See Label
For Use With (Application) Kinase Assay
Formulation Proprietary buffer with no preservative
Gene ID (Entrez) 24208
Name Rat Androgen Receptor, Ligand Binding Domain, GST Tag, His Tag
Purification Method Purified
Quantity 2 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AIS; Andr; androgen nuclear receptor variant 2; androgen receptor; androgen receptor (Testicular feminization), same as Tfm; androgen receptor splice variant 4b; Ar; AR8; AR-A; AW320017; DHTR; dihydrotestosterone receptor; HUMARA; HYSP1; KD; NR3C4; Nuclear receptor subfamily 3 group C member 4; RP11-383C12.1; SBMA; SMAX1; TFM; Variant AR45
Common Name Androgen Receptor
Gene Symbol AR
Protein Length 606-902
Product Type Protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Protein Tag GST-tag, His-tag
Sequence 606-902
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Growth Factors & Receptors
Protein Form Ligand Binding Domain, Recombinant
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.