Learn More
Medchemexpress LLC HY-N0191 100mg Medchemexpress, Andrographolide CAS:5508-58-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0191100MG
Encompass_Preferred
Medchemexpress, HY-N0191 100mg Andrographolide CAS:5508-58-7 Andrographolide is a NF-κB inhibitor, which inhibits NF-κB activation through covalent modification of a cysteine residue on p50 in endothelial cells without affecting IκBα degradation or p50/p65 nuclear translocation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3457