Learn More
Braintree Scientific Androit Heat Therapy Pump 120v
SDP
Catalog No. 501954941
Encompass
Heat Therapy Pump - The safest and most accurate localized heat therapy system available. Bright and easy-to-read display provides continuous real-time temperature readings. Digital controller uses proprietary software to manage users prescribed set-point temperature to within 1F. Three temperature safety limits guard against possibility of overheating and Hi Limit Switches can be tested without taking unit apart! Specifications: Set-point range: 75F (Off) to 107F, Set-point accuracy: .5F, >Flow rate: 15gph, Reservoir: 50 oz (1,500 ml), Approvals: uL 2601, CSA C22.2 No. 601-1 1DF, Mercury free, Uses tap water **Sold for research purposes only and not for human use**
Catalog No. 50-195-4941 Supplier Braintree Scientific Supplier No. HTP1500
 Restricted Item
Add to Cart
Add to Cart
Heat Therapy Pump - The safest and most accurate localized heat therapy system available. Bright and easy-to-read display provides continuous real-time temperature readings. Digital controller uses proprietary software to manage users prescribed set-point temperature to within 1F. Three temperature safety limits guard against possibility of overheating and Hi Limit Switches can be tested without taking unit apart! Specifications: Set-point range: 75F (Off) to 107F, Set-point accuracy: .5F, >Flow rate: 15gph, Reservoir: 50 oz (1,500 ml), Approvals: uL 2601, CSA C22.2 No. 601-1 1DF, Mercury free, Uses tap water **Sold for research purposes only and not for human use**