Learn More
Medchemexpress LLC HY-N1912 1mg Medchemexpress, Andropanolide CAS:869807-57-8 Purity:>98%
SDP
Catalog No. 501933858
Encompass_Preferred
Medchemexpress, HY-N1912 1mg Andropanolide CAS:869807-57-8 Andrographolide (Andro) is a small antagonist for NF-κB activation by covalent modifying reduced cysteine 62 of p50. Andrographolide is a bicyclic diterpenoid lactone mainly produced from the plant Andrographis (Andrographis paniculate).Andrographolide suppresses the activation of NF-κB in stimulated endothelial cells, which reduces the expression of cell adhesion molecule E-selectin and prevents E-selectin-mediated leukocyte adhesion, but has no effect on IκBα degradation, p50 and p65 nuclear translocation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3858 Supplier Medchemexpress LLC Supplier No. HYN19121MG
Medchemexpress, HY-N1912 1mg Andropanolide CAS:869807-57-8 Andrographolide (Andro) is a small antagonist for NF-κB activation by covalent modifying reduced cysteine 62 of p50. Andrographolide is a bicyclic diterpenoid lactone mainly produced from the plant Andrographis (Andrographis paniculate).Andrographolide suppresses the activation of NF-κB in stimulated endothelial cells, which reduces the expression of cell adhesion molecule E-selectin and prevents E-selectin-mediated leukocyte adhesion, but has no effect on IκBα degradation, p50 and p65 nuclear translocation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.