Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- | 115375-60-5 | 99.1% | C22H29F3O5S | 250 MG
SDP

Catalog No. 5000358688
Encompass_Preferred

This product is a drug intermediate used for the synthesis of various active compounds. It is supplied as a white to light yellow solid with high purity.

  • Purity of 99.09% (HPLC)
  • CAS number: 115375-60-5
  • Molecular formula: C22H29F3O5S
  • Molecular weight: 462.52
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-58688 Supplier Medchemexpress LLC Supplier No. 250MGHY75624
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a drug intermediate used for the synthesis of various active compounds. It is supplied as a white to light yellow solid with high purity.

  • Purity of 99.09% (HPLC)
  • CAS number: 115375-60-5
  • Molecular formula: C22H29F3O5S
  • Molecular weight: 462.52
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.