Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- | 115375-60-5 | 99.1% | C22H29F3O5S | 5 MG
SDP

Catalog No. 5000433775
Encompass_Preferred

This product, with CAS number 115375-60-5, is a high-purity (99.1%) chemical compound, Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)-. It functions as a versatile drug intermediate, crucial for the synthesis of various active compounds.

  • Chemical name: Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)-
  • Cas number: 115375-60-5
  • Molecular formula: C22H29F3O5S
  • Molecular weight: 462.52
  • Purity: 99.1%
  • Used as a drug intermediate for synthesis of various active compounds

Catalog No. 50-004-33775 Supplier Medchemexpress LLC Supplier No. HY756245MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product, with CAS number 115375-60-5, is a high-purity (99.1%) chemical compound, Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)-. It functions as a versatile drug intermediate, crucial for the synthesis of various active compounds.

  • Chemical name: Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)-
  • Cas number: 115375-60-5
  • Molecular formula: C22H29F3O5S
  • Molecular weight: 462.52
  • Purity: 99.1%
  • Used as a drug intermediate for synthesis of various active compounds

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.