Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- | 115375-60-5 | 99.1% | C22H29F3O5S | 50 MG
SDP

Catalog No. 5000433756
Encompass_Preferred

Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.

  • Used as a drug intermediate
  • Molecular weight: 462.52
  • Appearance: White to light yellow solid
  • Storage for powder: -20°C for 3 years; 4°C for 2 years
  • Storage for in-solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-33756 Supplier Medchemexpress LLC Supplier No. HY7562450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)- is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.

  • Used as a drug intermediate
  • Molecular weight: 462.52
  • Appearance: White to light yellow solid
  • Storage for powder: -20°C for 3 years; 4°C for 2 years
  • Storage for in-solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.