Learn More
Biomatik Corporation Androstenediol (AED) ELISA Kit
Supplier:  Biomatik Corporation EKU02345
Encompass_Preferred
Androstenediol (AED) ELISA Kit for detecting AED in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10000pg/ml. Sensitivity: 53.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 5-AED; 5AED; 5-Androstenediol; Androst-5-Ene-3Beta,17Beta-Diol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9137