Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Androstenedione Competitive ELISA Kit

Catalog No. EIAAND
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAAND 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAAND Supplier Invitrogen™ Supplier No. EIAAND
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The multispecies Androstenedione ELISA quantitates androstenedione in serum, plasma, and saliva, or urine. Principle of the method: The Androstenedione Competitive ELISA research-use-only kit is designed to quantitatively measure androstenedione independent of species. An adrostenedione standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with donkey anti-sheep IgG antibodies. Rabbit polyclonal antibodies and androstenedione-peroxidase conjugate is added to the wells. The androstenedione conjugate and any androstenedione in the sample will compete to bind to the rabbit androstenedione antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound testosteron-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of androstenedione present in the sample. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Androstenedione is a steroid hormone also known as androst-4-ene-3,17-dione. It is synthesized from dehydroepiandrosterone (DHEA) or 17-hydroxyprogesterone. By itself, androstenedione is a weak androgen though it is a precursor for biosynthesis of stronger androgens, such as testosterone and estrogens. Therefore, androstenedione is classified as a controlled drug and a supplement to enhance athletic performance and body energy.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 5-1200 pg/mL
Assay Sensitivity 2.3 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Chromone
Sample Type Plasma, Saliva, Serum, Urine
Target Species Chemical
For Use With (Equipment) Absorbance Microplate Reader
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers, Standard, Antibody, HRP-Conjugated Detection Antibody, Assay Buffer Concentrate, Wash Buffer, TMB Substrate, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Saliva 50 μL, Serum 50 μL, Urine 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target Androstenedione
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.