Learn More
Drg International Inc Androstenedione Elisa
Supplier:  Drg International Inc EIA3265
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of androstenedione in serum and EDTA plasma (Do not use Heparin or Citrate plasma). The steroid hormone Androstenedione is one of the main androgens, besides Testosterone and Dehydroepiandrosterone. The DRG Androstenedione ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [rabbit] antibody directed towards an antigenic site of the Androstenedione molecule. Endogenous Androstenedione of a patient sample competes with an Androstenedione-horseradish peroxidase conjugate for binding to the coated antibody.
Catalog No. 50-194-7553