Learn More
Arbor Assays Androstenedione ELISA Kit, (5 Plate)
SDP
Supplier:  Arbor Assays K070H5
Encompass_Preferred
The DetectX Androstenedione ELISA Kits quantitatively measure androstenedione in urine, saliva, or extracted serum and plasma.
Catalog No. 50-227-8511