Learn More
Cayman Chemical Androsterone 1g
SDP
Catalog No. 502971185
Encompass_Preferred
A covalent inhibitor of PME-1 (IC50 4.2 nM) selectively inhibits PME-1 over 50 other serine hydrolases in MDA-MB-231 and HEK293T cell lysates at 10 M decreases proliferation of Ishikawa cells and migration of ECC-1 endometrial carcinoma cells when used at a concentration of 50 nM
Catalog No. 50-297-1185 Supplier Cayman Chemical Supplier No. 15872 1G
A covalent inhibitor of PME-1 (IC50 4.2 nM) selectively inhibits PME-1 over 50 other serine hydrolases in MDA-MB-231 and HEK293T cell lysates at 10 M decreases proliferation of Ishikawa cells and migration of ECC-1 endometrial carcinoma cells when used at a concentration of 50 nM