Learn More
Andwin Scientific Kettle Mantles for PYREX™ Reaction Kettles
SDP
Catalog No. 1184715E
Click to view available options
For Use With (Equipment):
11-847B kettles
11-847E Kettles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-847-15E 11-847E Kettles
11-847-15B 11-847B kettles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-847-15E Supplier Andwin Scientific Supplier No. TM 580
Lower sections moldblown for breakage resistance
  • Heavy pressed covers have four openings to accommodate interchangeable standard taper joints, allowing quick assembly with condensers, stirrers and funnels
  • Glass kettles designed for organic reactions involving viscous materials
  • Wide mouth for easy cleaning of interior
  • Finely ground flanges for tight seal
  • 500 and 1000mL sizes have interchangeable covers with four standard taper 24/40 joints; 2000, 3000, 4000mL sizes have one standard taper 34/45 and three standard taper 24/40 joints
Certifications
CSA certified
†Mantles available for other types of reaction kettles. Call 1-800-766-7000 for details.
Specifications
For Use With (Equipment) 11-847E Kettles
Product Type Mantle