Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific 100-Slide Capacity Microscope Slide Storage Boxes
SDP

Catalog No. 1437288
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Red
White
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-372-88 White
14-372-85 Red
14-372-86 Blue
14-372-87 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-372-88 Supplier Andwin Scientific Supplier No. 880120
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for safe, long-term slide storage

  • Constructed from polystyrene
  • 100-Slide capacity
  • Corrosion-resistant hinges and clasp
  • Cork-lined bottoms
  • Individually numbered slide positions
  • Available in four colors

Specifications

Slide Capacity 100
Color White
Device Holds 100
For Use With (Equipment) Microscope Slide
Includes Cork lined bottom
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.