Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific 25-Slide Capacity Microscope Slide Storage Box
SDP

Catalog No. 1437289
Encompass_Preferred
Change view
Click to view available options
Slide Capacity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
14-372-89 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-372-89 Supplier Andwin Scientific Supplier No. 880145
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Constructed from durable polystyrene

  •  25-slide capacity
  • Cork-lined bottom
Note: Actual product does not have hinges as shown in image above; rather it consists of two pieces - box and lid.

Specifications

Slide Capacity 25
Color Blue
For Use With (Equipment) Microscope Slide
Includes Cork lined bottom
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.