Learn More
Andwin Scientific Boritex™ Urine Stabilization System
SDP
Catalog No. 1437356
Encompass_Preferred
Change view
Click to view available options
Closure Color:
Yellow
Catalog No. Closure Color
14-373-56 Yellow
1 options
This item is not returnable. View return policy
Catalog No. 14-373-56 Supplier Andwin Scientific Supplier No. 700090
This item is not returnable. View return policy
Includes
50mg Boritex™ tablet, Tube, Screw cap
Preserves critical cells and potential bacterial pathogens for up to 96 hours
  • Flat bottom polypropylene vial with yellow screw cap
  • Nonsterile
Specifications
Product Type Urine Stabilization System
Closure Color Yellow
Closure Type Screw Cap
Sterility Non-sterile
Bottom Shape Flat
For Use With (Application) For remote collection sites
Includes 12 mL Conical tube, 50 mg Boritex tablet, tube, screw cap