Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific Lazy L-shaped Spreader
SDP

Catalog No. 1437376
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-373-76 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-373-76 Supplier Andwin Scientific Supplier No. 900500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L–shaped for a smooth and even sample distribution.

  • Polystyrene spreader
  • Durable disposable and non–breakable
  • Shape reduces the likelihood of the spreader digging into the agar
  • Pre–sterilized
  • Blue

Specifications

Disposable Single-use
Autoclavable Not Autoclavable
Shape L-Shape
Sterility Sterile
Color Blue
Material Polystyrene
Product Type Cell Spreader
For Use With (Application) Allows for a smooth and even sample distribution, reducing the likelihood of the spreader digging into the agar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.