Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific Safetex™ Cytology Fixative
SDP

Catalog No. 1437246
Encompass_Preferred
Change view
Click to view available options
Product Type:
Fixative
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14-372-46 Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-372-46 Supplier Andwin Scientific Supplier No. 930010C12
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Fixative
Quantity 29.6mL
Formulation Water and Alcohol soluble Polyethylene Glycol
For Use With (Application) Industrial use
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.