Learn More
Medchemexpress LLC Anecortave acetate 10mg | 7753-60-8 | 386.5 g/mol | C23H30O5 | 10 MG
SDP
Catalog No. 5000227660
Encompass_Preferred
Anecortave acetate is a steroidal angiostatic agent used in research on ocular neovascularization and age-related macular degeneration. It is supplied as a high-purity research reagent with supporting analytical documentation and handling guidance.
  • High reported purity, suitable for research use.
  • Molecular weight 386.5 g/mol and formula C23H30O5.
  • Available in a 10 mg research package.
  • Analytical documentation available, including COA, HNMR, and LCMS.
  • Used in studies of angiogenesis and retinal neovascular disorders.
Catalog No. 50-002-27660 Supplier Medchemexpress LLC Supplier No. HY11686810MG
Anecortave acetate is a steroidal angiostatic agent used in research on ocular neovascularization and age-related macular degeneration. It is supplied as a high-purity research reagent with supporting analytical documentation and handling guidance.
  • High reported purity, suitable for research use.
  • Molecular weight 386.5 g/mol and formula C23H30O5.
  • Available in a 10 mg research package.
  • Analytical documentation available, including COA, HNMR, and LCMS.
  • Used in studies of angiogenesis and retinal neovascular disorders.