Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anecortave acetate 5mg | 7753-60-8 | 386.48 g/mol | C23H30O5 | 5 MG
SDP

Catalog No. 5000227662
Encompass_Preferred

Anecortave acetate is an angiostatic research compound that inhibits pathological retinal neovascularization. Supplied as a high-purity solid for laboratory and preclinical studies, it is soluble in methanol and requires cold storage to maintain stability.

  • High purity (99.81%).
  • Solid form for handling and storage.
  • Molecular formula C23H30O5 and molecular weight 386.48 g/mol.
  • Soluble in methanol: 1 mg/mL; may require ultrasonic.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research sizes, including 5 mg.

Catalog No. 50-002-27662 Supplier Medchemexpress LLC Supplier No. HY1168685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anecortave acetate is an angiostatic research compound that inhibits pathological retinal neovascularization. Supplied as a high-purity solid for laboratory and preclinical studies, it is soluble in methanol and requires cold storage to maintain stability.

  • High purity (99.81%).
  • Solid form for handling and storage.
  • Molecular formula C23H30O5 and molecular weight 386.48 g/mol.
  • Soluble in methanol: 1 mg/mL; may require ultrasonic.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research sizes, including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.