Learn More
Medchemexpress LLC HY-N0813 5mg Medchemexpress, Anemarsaponin E CAS:136565-73-6 Purity:>98%
SDP
Catalog No. 501933728
Encompass_Preferred
Medchemexpress, HY-N0813 5mg Anemarsaponin E CAS:136565-73-6 Anemarsaponin E is extracted from Anemarrhena asphodeloides Bunge and has anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3728 Supplier Medchemexpress LLC Supplier No. HYN08135MG
Medchemexpress, HY-N0813 5mg Anemarsaponin E CAS:136565-73-6 Anemarsaponin E is extracted from Anemarrhena asphodeloides Bunge and has anti-inflammatory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.