Learn More
Cole-Parmer TSI 9515 VelociCalc Air Velocity Meter, Remote Probe
Supplier:  Cole-Parmer 1027025
Encompass
These air velocity meters are ideal for measuring face velocity in fume hoods, spray booths, ventilation systems, or IAQ applications.
Catalog No. 50-633-404