Learn More
Cole-Parmer Extech 407113 Heavy-Duty CFM Metal Vane Thermoanemometer
Supplier:  Cole-Parmer 1020007
Encompass
Meter displays air volume(CFM), air velocity, and temperature simultaneously on the large LCD display. Other features include Min/Max, data hold, record/recall, and auto shut-off. Use optional data module 10200-20 to store up to 8000 data points. Order optional RS-232 interface to download data directly to a PC. Note: not designed for heat applications.
Catalog No. 50-992-909