Learn More
Cole-Parmer Testo 0560 4053 Stick Thermoanemometer; in metric only
Supplier:  Cole-Parmer 3746600
Encompass
This anemometer measures air velocity, temperature and air volume. Miniature hot-wire sensor measures parameters in hard-to-reach areas. Rotating cap protects the sensor during storage. Meter features a single push button control, and includes a rubber duct mount holder for convenient duct measurements. Auto-off function conserves battery life. Whats included: pocket clip , rubber duct holder, and three AAA batteries
Catalog No. NC9948933