Learn More
Selleck Chemical LLC Anemoside B4 S9081-1MG
SDP
Catalog No. 5000025439
Encompass_Preferred
Anemoside B4 (Pulchinenoside B4 Chinensioside A Pulchinenoside C Pulsatilla saponin B4) extracted from the herb of Pulsatilla chinensis (Bge ) Regel is a potential antiviral constituent and inhibits the secretion of IL-10
Catalog No. 50-000-25439 Supplier Selleck Chemical LLC Supplier No. S90811MG
Anemoside B4 (Pulchinenoside B4 Chinensioside A Pulchinenoside C Pulsatilla saponin B4) extracted from the herb of Pulsatilla chinensis (Bge ) Regel is a potential antiviral constituent and inhibits the secretion of IL-10