Learn More
TARGETMOL CHEMICALS INC Aneratrigine hydrochloride 1MG
SDP
Catalog No. 502876592
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aneratrigine hydrochloride is a sodium channel protein type 9 subunit blocker that may be used for the prevention or treatment of sodium channel blocker-related diseases. Purity 99.39%
Catalog No. 50-287-6592 Supplier TARGETMOL CHEMICALS INC Supplier No. T798351MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aneratrigine hydrochloride is a sodium channel protein type 9 subunit blocker that may be used for the prevention or treatment of sodium channel blocker-related diseases. Purity 99.39%