Learn More
Med Vet International Anesthesia Breathing Tube 36"L, J0249H
Catalog No. 5000582079
Encompass_Preferred
Corrugated black rubber conductive breathing tubes with natural rubber 22mm cuff inserts.
Catalog No. 50-005-82079 Supplier Med Vet International Supplier No. JOR249H
Corrugated black rubber conductive breathing tubes with natural rubber 22mm cuff inserts.