Learn More
Med Vet International Anesthesia Chamber with Dividers, J0265
Catalog No. 5000585544
Encompass_Preferred
The Lexan anesthesia chamber provides a simple method of induction. Chamber induction reduces physical restraint which minimizes stress and reduces the chance of injury to the operator and animal. The divider system allows for up to four different chamber sizes within one unit: 18" x 9" x 9"; 12" x 9" x 9"; 6" x 9" x 9". Unit weighs 10 lbs.
Catalog No. 50-005-85544 Supplier Med Vet International Supplier No. JOR265
The Lexan anesthesia chamber provides a simple method of induction. Chamber induction reduces physical restraint which minimizes stress and reduces the chance of injury to the operator and animal. The divider system allows for up to four different chamber sizes within one unit: 18" x 9" x 9"; 12" x 9" x 9"; 6" x 9" x 9". Unit weighs 10 lbs.