Learn More
Med Vet International Anesthesia Circuit Cleaning Brush, Silicone, 3.5mm, J1524, Each
Catalog No. 5000583713
Encompass_Preferred
Finally a cleaning brush that can clean the inside of an anesthesia circuit reaching inside every recess for a thorough cleaning. It is difficult and expensive to dispose of anesthesia circuit after just one single use.  Vet clinics have tried numerous methods to safely clean and sanitize anesthesia circuits between uses. It is not an easy task. • Works well in corrugated tube with a tight fit • Total reach is 36", then enter the opposite end to finish the job • Please note: Easy assembled cleaning rod screws onto soft plastic brush • Too large for pediatric smaller tubing • Too large for 19mm scavenger tubing
Catalog No. 50-005-83713 Supplier Med Vet International Supplier No. JOR1524
Finally a cleaning brush that can clean the inside of an anesthesia circuit reaching inside every recess for a thorough cleaning. It is difficult and expensive to dispose of anesthesia circuit after just one single use.  Vet clinics have tried numerous methods to safely clean and sanitize anesthesia circuits between uses. It is not an easy task. • Works well in corrugated tube with a tight fit • Total reach is 36", then enter the opposite end to finish the job • Please note: Easy assembled cleaning rod screws onto soft plastic brush • Too large for pediatric smaller tubing • Too large for 19mm scavenger tubing