Learn More
World Precision Instrument Anesthesia Connection Kit
SDP
Catalog No. 502538849
Encompass
Anesthesia Connection Kit
This kit is required for integrating third party equipment into an EZ Ansthesia system. It includes EZ-2SF Female Quick Disconnect with 6 ft of 0.25" ID clear PVC tubing with clamps.
Catalog No. 50-253-8849 Supplier World Precision Instrument Supplier No. EZ1130
Anesthesia Connection Kit
This kit is required for integrating third party equipment into an EZ Ansthesia system. It includes EZ-2SF Female Quick Disconnect with 6 ft of 0.25" ID clear PVC tubing with clamps.