Learn More
Med Vet International Anesthesia Mask Canine 5" Opening, J0298C
Catalog No. 5000583089
Encompass_Preferred
Anesthesia Masks with low dead space, clear plexiglass masks with black rubber diaphragm.
Catalog No. 50-005-83089 Supplier Med Vet International Supplier No. JOR298C
Anesthesia Masks with low dead space, clear plexiglass masks with black rubber diaphragm.