Learn More
Med Vet International Anesthesia Mask Feline, 3.5" Opening, J0298F
Catalog No. 5000581499
Encompass_Preferred
Anesthesia Masks with low dead space, clear plexiglass masks with black rubber diaphragm.15 mm i.d. on the end where the endotracheal tube connects
Catalog No. 50-005-81499 Supplier Med Vet International Supplier No. JOR298F
Anesthesia Masks with low dead space, clear plexiglass masks with black rubber diaphragm.15 mm i.d. on the end where the endotracheal tube connects